Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-heptylammonium iodide, 99%

$42.96 - $292.46

Chemical Identifiers

Chemical Name or Material Tetra-n-heptylammonium iodide
CAS 3535-83-9
Molecular Formula [CH3(CH2)6]4NI
Formula Weight 537.7
MDL Number MFCD00041984
Beilstein 4167115
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1664206

 
alfa aesar™
A1664206
5g Each for $42.96

AAA1664214

 
alfa aesar™
A1664214
25g Each for $128.07

AAA1664222

 
alfa aesar™
A1664222
100g Each for $292.46
Description & Specifications

Specifications

Formula Weight 537.7
Chemical Name or Material Tetra-n-heptylammonium iodide
TSCA TSCA
Assay 99%
Molecular Formula [CH3(CH2)6]4NI
MDL Number MFCD00041984
CAS 3535-83-9
Beilstein 4167115
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 122° to 124°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.