Learn More
Tetra-n-heptylammonium bromide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1521822
Description
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Tetra-n-heptylammonium bromide | |
87°C to 89°C | |
MFCD00011861 | |
Hygroscopic | |
Soluble in water and methanol. | |
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-] | |
490.699 | |
490.7 |
4368-51-8 | |
C28H60BrN | |
100 g | |
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1- | |
YQIVQBMEBZGFBY-UHFFFAOYSA-M | |
tetraheptylazanium;bromide | |
78073 | |
99% |
Chemical Identifiers
4368-51-8 | |
490.699 | |
YQIVQBMEBZGFBY-UHFFFAOYSA-M | |
78073 | |
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-] |
C28H60BrN | |
MFCD00011861 | |
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1- | |
tetraheptylazanium;bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-459-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only