Please login to your online account to display your discounted pricing

Tetra-n-butyltin, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetra-n-butyltin
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C16H36Sn
Linear Formula [CH3(CH2)3]4Sn
Formula Weight 347.14
MDL Number MFCD00009422
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC137985000

  SDS
thermo fisher scientific
AC137985000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 347.14
Color Clear colorless to straw
Quantity 500g
Density 1.057g/mL
Chemical Name or Material Tetra-n-butyltin
Linear Formula [CH3(CH2)3]4Sn
CAS 1461-25-2
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C16H36Sn
MDL Number MFCD00009422
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.