missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetra-n-butylphosphonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041718K2

 View more versions of this product

Catalog No. AA41718K2



Description

Description

Tetra-n-butylphosphonium hydroxide is used for the extraction of wheat straw and dissolution of wood. It is also used to produce hydroxide ion or proton conductive ionic liquids by mixing it with benzimidazole. It can be used as a phase transfer catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetra-n-butylphosphonium hydroxide
Colorless to Yellow
C16H37OP
[CH3(CH2)3]4POH
1 L
tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
DFQPZDGUFQJANM-UHFFFAOYSA-M
tetrabutylphosphanium;hydroxide
84487
40% w/w aq. soln.
14518-69-5
0.989 g/mL
1.412
MFCD00068456
UN3267
Miscible with water.
[OH-].CCCC[P+](CCCC)(CCCC)CCCC
276.45
276.45
Liquid

Chemical Identifiers

14518-69-5
276.45
DFQPZDGUFQJANM-UHFFFAOYSA-M
84487
[OH-].CCCC[P+](CCCC)(CCCC)CCCC
C16H37OP
MFCD00068456
tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
tetrabutylphosphanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

EINECSNumber : 238-528-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only