Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butyllead, 95%

Chemical Identifiers

Chemical Name or Material Tetra-n-butyllead
CAS 1920-90-7
Molecular Formula C16H36Pb
Linear Formula [CH3(CH2)3]4Pb
Formula Weight 435.66
MDL Number MFCD00015226
UN Number UN3282
Synonym Lead tetrabutyl; Tetra-n-butylplumbane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA5712903

  SDS
alfa aesar™
5712903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA5712906

  SDS
alfa aesar™
5712906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 435.66
Solubility Information Insoluble in water
Linear Formula [CH3(CH2)3]4Pb
Quantity 1g
Synonym Lead tetrabutyl; Tetra-n-butylplumbane
MDL Number MFCD00015226
CAS 1920-90-7
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.
Color Colorless to yellow
Sensitivity Air & Light Sensitive
Health Hazard 2 GHS H Statement
H300-H310-H330-H360-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flash Point 140°C (284°F)
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Physical Form Liquid, May Include Precipitate
Chemical Name or Material Tetra-n-butyllead
UN Number UN3282
Boiling Point 140°C (1mmHg)
Molecular Formula C16H36Pb
Density 1.324g/mL
Refractive Index 1.5119
Assay 95%
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H360-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.