missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butyllead, 95%

Manufacturer:  Alfa Aesar™ 5712906

 View more versions of this product

Catalog No. AA5712906



Specifications

Specifications

Tetra-n-butyllead
1920-90-7
140°C (1mmHg)
140°C (284°F)
1.5119
[CH3(CH2)3]4Pb
UN3282
Insoluble in water
CCCC[Pb](CCCC)(CCCC)CCCC
435.664
435.66
95%
5g
1.324g/mL
Colorless to Yellow
Air & Light Sensitive
C16H36Pb
MFCD00015226
plumbane, tetrabutyl, tetra-n-butyllead, lead tetra-n-butyl, tetra-n-butyl lead, lead tetrabutyl, phthalocyanine, ferric, kdqhjgwpoqncmi-uhfffaoysa
KDQHJGWPOQNCMI-UHFFFAOYSA-N
tetrabutylplumbane
15978
Liquid, May Include Precipitate

Chemical Identifiers

1920-90-7
435.664
KDQHJGWPOQNCMI-UHFFFAOYSA-N
15978
CCCC[Pb](CCCC)(CCCC)CCCC
C16H36Pb
MFCD00015226
plumbane, tetrabutyl, tetra-n-butyllead, lead tetra-n-butyl, tetra-n-butyl lead, lead tetrabutyl, phthalocyanine, ferric, kdqhjgwpoqncmi-uhfffaoysa
tetrabutylplumbane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H360-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.

TSCA : TSCA

SDS Documents