Learn More
Tetra-n-butylammonium tribromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01625814
Description
Tetra-n-butylammonium tribromide is used as a phase transfer catalyst in organic synthesis as well as a mild brominating agent. It acts as a solid source of bromine. It is useful for the preparation of vicinal dibromides from alkenes and alkynes and alfa-bromo acetals. Further, it is utilized to generate hydrogen bromide, which is used as an efficient chemoselective reagent for acetalization of carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-butylammonium tribromide | |
74°C to 76°C | |
C48H108Br3N3 | |
MFCD00012110 | |
UN3261 | |
tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide | |
SFLXUZPXEWWQNH-UHFFFAOYSA-K | |
tetrabutylazanium;tribromide | |
23500184 | |
98% |
38932-80-8 | |
Yellow | |
[CH3(CH2)3]4NBr3 | |
25 g | |
3746114 | |
Insoluble in water. | |
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-] | |
967.125 | |
482.18 | |
Crystalline |
Chemical Identifiers
38932-80-8 | |
967.125 | |
SFLXUZPXEWWQNH-UHFFFAOYSA-K | |
23500184 | |
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-] |
C48H108Br3N3 | |
MFCD00012110 | |
tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide | |
tetrabutylazanium;tribromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only