Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium tetraphenylborate, 99+%

$24.11 - $24.11

Chemical Identifiers

Chemical Name or Material Tetra-n-butylammonium tetraphenylborate
CAS 15522-59-5
Molecular Formula C40H56BN
Linear Formula [CH3(CH2)3]4NB(C6H5)4
Formula Weight 561.7
MDL Number MFCD00011749
Beilstein 3857215
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0098503

  SDS
alfa aesar™
L0098503
1g Each for $24.11
Description & Specifications

Specifications

Formula Weight 561.7
Chemical Name or Material Tetra-n-butylammonium tetraphenylborate
Molecular Formula C40H56BN
MDL Number MFCD00011749
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 232° to 235°C
Quantity 1g
Assay ≥99%
Beilstein 3857215
Linear Formula [CH3(CH2)3]4NB(C6H5)4
CAS 15522-59-5
Health Hazard 1 Warning

Applications
Tetra-n-butylammonium tetraphenylborate, is used as a supporting electrolyte and standard for cell calibration. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in pyridine, acetone, water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.