Learn More
Tetra-n-butylammonium sulfate, 50% w/w aq. soln., Thermo Scientific Chemicals
Phase transfer catalyst, neutral pH
Supplier: Thermo Scientific Chemicals 041723K4
Description
Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-butylammonium sulfate | |
1.01 | |
Unpleasant | |
1.417 | |
2.5 L | |
Fully miscible. | |
[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC | |
581.00 | |
580.99 | |
Liquid |
2472-88-0 | |
84°C | |
C32H72N2O4S | |
MFCD00216632 | |
bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate | |
ZXUCBXRTRRIBSO-UHFFFAOYSA-L | |
tetrabutylazanium;sulfate | |
11433187 | |
50% w/w aq. soln. |
Chemical Identifiers
2472-88-0 | |
581.00 | |
ZXUCBXRTRRIBSO-UHFFFAOYSA-L | |
11433187 | |
[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC |
C32H72N2O4S | |
MFCD00216632 | |
bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate | |
tetrabutylazanium;sulfate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-593-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only