missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium phosphate, 99+%

Used as a counter-ion in ion-pair chromatography of water-soluble vitamins

Manufacturer:  Alfa Aesar™ 1926118

 View more versions of this product

Catalog No. AA1926118



Specifications

Specifications

Tetra-n-butylammonium phosphate
151°C to 154°C
5574-97-0
[CH3(CH2)3]4NH2PO4
5196532
Soluble in acetonitrile, ethanol, water
CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)(O)[O-]
339.457
339.46
≥99%
White
50g
C16H38NO4P
MFCD00064526
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
dihydrogen phosphate;tetrabutylazanium
2735142
Crystalline

Chemical Identifiers

5574-97-0
339.457
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
2735142
CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)(O)[O-]
C16H38NO4P
MFCD00064526
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
dihydrogen phosphate;tetrabutylazanium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents