Learn More
Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04674604
Description
Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-butylammonium phosphate non-UV | |
151°C to 154°C | |
C16H38NO4P | |
MFCD00064526 | |
5196532 | |
It is soluble in water. | |
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
339.46 | |
339.46 | |
Crystalline |
5574-97-0 | |
White | |
[CH3(CH2)3]4NH2PO4 | |
2 g | |
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
dihydrogen phosphate;tetrabutylazanium | |
2735142 | |
98% |
Chemical Identifiers
5574-97-0 | |
339.46 | |
ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
2735142 | |
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
C16H38NO4P | |
MFCD00064526 | |
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
dihydrogen phosphate;tetrabutylazanium |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
H500
EINECSNumber : 226-947-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only