missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium perchlorate, electrochemical grade

Suitable for use as a polarographic supporting electrolyte

Manufacturer:  Alfa Aesar™ 4399930

 View more versions of this product

Catalog No. AA4399930



Specifications

Specifications

Tetra-n-butylammonium perchlorate
White
250g
C16H36ClNO4
MFCD00038722
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
KBLZDCFTQSIIOH-UHFFFAOYSA-M
tetrabutylazanium;perchlorate
74723
Crystalline
Hygroscopic
210°C to 217°C
1923-70-2
[CH3(CH2)3]4NClO4
UN1481
Soluble in ethanol; Very slightly soluble in water
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O
341.917
341.92
Electrochemical

Chemical Identifiers

1923-70-2
341.917
KBLZDCFTQSIIOH-UHFFFAOYSA-M
74723
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O
C16H36ClNO4
MFCD00038722
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
tetrabutylazanium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents