missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetra-n-butylammonium perchlorate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03080188

 View more versions of this product

Catalog No. AA3080188



Description

Description

Tetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetra-n-butylammonium perchlorate
210°C to 217°C
C16H36ClNO4
MFCD00038722
UN1481
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
KBLZDCFTQSIIOH-UHFFFAOYSA-M
tetrabutylazanium;perchlorate
74723
≥99%
1923-70-2
White
[CH3(CH2)3]4NClO4
2.5 g
Hygroscopic
Soluble in acetonitrile and ethanol. Slightly soluble in water.
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O
341.917
341.92
Crystalline

Chemical Identifiers

1923-70-2
341.917
KBLZDCFTQSIIOH-UHFFFAOYSA-M
74723
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O
C16H36ClNO4
MFCD00038722
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
tetrabutylazanium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 217-655-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only