Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium L-lactate, 70% w/w aq. soln.

$27.43 - $27.43

Chemical Identifiers

Chemical Name or Material Tetra-n-butylammonium L-lactate
Molecular Formula [CH3(CH2)3]4N[CH3CH(OH)CO2]
Formula Weight 331.54
MDL Number MFCD00216638
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43035AA

  SDS
alfa aesar™
43035AA
5mL Each for $27.43
Description & Specifications

Specifications

Formula Weight 331.54
Density 0.957g/mL
Physical Form Liquid
Health Hazard 1 Warning
Molecular Formula [CH3(CH2)3]4N[CH3CH(OH)CO2]
Quantity 5mL
Concentration 70% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium L-lactate
MDL Number MFCD00216638
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.