missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium iodotetrachloride, 95%

$76.19 - $265.24

Chemical Identifiers

CAS 15625-59-9
Molecular Formula C16H36Cl4IN
Molecular Weight (g/mol) 511.175
MDL Number MFCD00051851
InChI Key HBZJAVINJZUOEH-UHFFFAOYSA-N
Synonym tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide
PubChem CID 73555556
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0057406 View Documents Alfa Aesar™
L0057406
5g Each for $76.19
 
AAL0057414 View Documents Alfa Aesar™
L0057414
25g Each for $265.24
 
Specifications

Chemical Identifiers

15625-59-9
511.175
HBZJAVINJZUOEH-UHFFFAOYSA-N
73555556
C16H36Cl4IN
MFCD00051851
tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide
CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl

Specifications

135°C to 139°C
15625-59-9
MFCD00051851
HBZJAVINJZUOEH-UHFFFAOYSA-N
511.175
511.19
Tetra-n-butylammonium iodotetrachloride
5g
C16H36Cl4IN
tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide
CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl
73555556
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C