Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium hydroxide, Reagent Grade, 31% in methanol

As a basic nonaqueous titrant

Chemical Identifiers

Chemical Name or Material Tetra-n-butylammonium hydroxide
CAS 2052-49-5
Purity Grade Reagent Grade
Formula Weight 259.48g/mol
MDL Number MFCD00009425
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1626306

 
alfa aesar™
16263-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1626322

 
alfa aesar™
16263-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.830g/mL
Refractive Index 1.3775
Flash Point 13.9° (57°F)
Odor Alcohol-like
Concentration 31% in methanol
Quantity 5g
Chemical Name or Material Tetra-n-butylammonium hydroxide
CAS 2052-49-5
Purity Grade Reagent Grade
Physical Form Liquid
Formula Weight 259.48g/mol
MDL Number MFCD00009425
UN Number UN2924
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H300-H310-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H300-H310-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.