missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium hydroxide, 55% w/w aq. soln.

As a reagent for ion-pair extractions

Manufacturer:  Alfa Aesar™ 1619830

 View more versions of this product

Catalog No. AA1619830



Specifications

Specifications

Tetra-n-butylammonium hydroxide
1.435
Colorless to Yellow
14
C16H37NO
MFCD00009425
3571228
55% w/w aq. soln.
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
259.478
259.48
0.97g/mL
Air Sensitive
250g
2052-49-5
[CH3(CH2)3]4NOH
UN3267
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium;hydroxide
2723671
Liquid

Chemical Identifiers

2052-49-5
259.478
VDZOOKBUILJEDG-UHFFFAOYSA-M
2723671
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
C16H37NO
MFCD00009425
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
tetrabutylazanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

RTECSNumber : BS5425000

TSCA : TSCA

SDS Documents