missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium hydroxide, 1.0M aq. soln.

Manufacturer:  Alfa Aesar™ 46315AE

 View more versions of this product

Catalog No. AA46315AE



Specifications

Specifications

Tetra-n-butylammonium hydroxide
100mL
C16H37NO
MFCD00009425
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium;hydroxide
2723671
Liquid
Air Sensitive
2052-49-5
[CH3(CH2)3]4NOH
UN3267
1.0M aq. soln.
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
259.478
259.48

Chemical Identifiers

2052-49-5
259.478
VDZOOKBUILJEDG-UHFFFAOYSA-M
2723671
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
C16H37NO
MFCD00009425
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
tetrabutylazanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

RTECSNumber : BS5425000

TSCA : TSCA

SDS Documents