Please login to your online account to display your discounted pricing

Tetra-n-butylammonium hexafluorophosphate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  352580250

 View more versions of this product

Catalog No. AC352580250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder and/or Chunks
Formula Weight 387.44
Quantity 25g
Color White to off-white
Melting Point 242° to 247°C
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula [CH3(CH2)3]4N(PF6)
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Elemental Analysis 97.5% min.
Molecular Formula C16H36N·PF6
Health Hazard 1 Danger
Assay Percent Range 98%
Chemical Name or Material Tetra-n-butylammonium hexafluorophosphate
CAS 3109-63-5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Tetra-n-butylammonium hexafluorophosphate
Assay Percent Range 98%
Molecular Formula C16H36N·PF6
Linear Formula [CH3(CH2)3]4N(PF6)
Formula Weight 387.44
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents