missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium fluoride trihydrate, 98%

Manufacturer:  Alfa Aesar™ L1330318

 View more versions of this product

Catalog No. AAL1330318



Description

Description

Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections in vivo and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetra-n-butylammonium fluoride trihydrate
54°C to 60°C
87749-50-6
[CH3(CH2)3]4NF·3H2O
UN2811
tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]
315.514
315.52 (261.47 Anhydrous)
Odorless
50g
C16H42FNO3
MFCD00149981
3761900
VEPTXBCIDSFGBF-UHFFFAOYSA-M
tetrabutylazanium;fluoride;trihydrate
11726816
98%

Chemical Identifiers

87749-50-6
315.514
VEPTXBCIDSFGBF-UHFFFAOYSA-M
11726816
CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]
C16H42FNO3
MFCD00149981
tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
tetrabutylazanium;fluoride;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H318-H315
Fatal if swallowed.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Documents