Learn More
Tetra-n-butylammonium difluorotriphenylstannate, 97%, Thermo Scientific™
$77.58 - $287.74
Chemical Identifiers
CAS | 139353-88-1 |
---|---|
Molecular Formula | C34H51F2NSn |
Molecular Weight (g/mol) | 630.496 |
MDL Number | MFCD00192465 |
InChI Key | ODMXVCNGZGLSRS-UHFFFAOYSA-L |
Synonym | difluoro triphenyl stannanuide; tetrabutylammonium, difluorotriphenylstannanuide; tetrabutylammonium, tetra-n-butylammonium difluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate |
PubChem CID | 2733194 |
IUPAC Name | difluoro(triphenyl)stannanuide;tetrabutylazanium |
SMILES | CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2020503
|
Thermo Fisher Scientific
L2020503 |
1g |
Each for $77.58
|
|
AAL2020506
|
Thermo Fisher Scientific
L2020506 |
5g |
Each for $287.74
|
|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCan be regarded as a non-hygroscopic anhydrous equivalent of TBAF useful in the preparation of gem-difluorides and the alkylation of enol silyl ethers. For use in the fluorination of carbohydrates.
Solubility
Soluble in most organic solvents.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
139353-88-1 | |
630.496 | |
ODMXVCNGZGLSRS-UHFFFAOYSA-L | |
2733194 | |
CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F |
C34H51F2NSn | |
MFCD00192465 | |
difluoro triphenyl stannanuide; tetrabutylammonium, difluorotriphenylstannanuide; tetrabutylammonium, tetra-n-butylammonium difluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate | |
difluoro(triphenyl)stannanuide;tetrabutylazanium |
Specifications
139353-88-1 | |
C34H51F2NSn | |
UN3146 | |
difluoro triphenyl stannanuide; tetrabutylammonium, difluorotriphenylstannanuide; tetrabutylammonium, tetra-n-butylammonium difluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate | |
Soluble in most organic solvents. | |
ODMXVCNGZGLSRS-UHFFFAOYSA-L | |
difluoro(triphenyl)stannanuide;tetrabutylazanium | |
2733194 | |
97% |
188°C to 191°C | |
MFCD00192465 | |
8668854 | |
1g | |
Hygroscopic | |
CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F | |
630.496 | |
630.5 | |
Tetra-n-butylammonium difluorotriphenylstannate |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures