missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium dichromate, 99%

Manufacturer:  Alfa Aesar™ H5520514

 View more versions of this product

Catalog No. AAH5520514



Description

Description

Applications
Reagent for preparation of nitrophenol derivatives and quinones under aprotic reaction conditions, conversion of oximes to carbonyl compounds under microwave irradiation, catalyst precursor for catalytic gas phase dehydration of acetic acid to ketene, crosslinking agent.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Tetra-n-butylammonium dichromate
25g
C32H72Cr2N2O7
MFCD00013104
bis tetrabutylammonium dichromate, tbadc, tetrabutylammonium dichromate, bis tetrabutyl ammonium dichromate, acmc-1avk0, tetra-n-butylammonium dichromate, bis tetrabutylammonium ion chromate
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
700.927
700.92
139°C to 143°C
56660-19-6
([CH3(CH2)3]4N)2(Cr2O7)
UN1479
MMKQSIQNDMIVIN-UHFFFAOYSA-N
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;tetrabutylazanium
11767684
99%

Chemical Identifiers

56660-19-6
700.927
MMKQSIQNDMIVIN-UHFFFAOYSA-N
11767684
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
C32H72Cr2N2O7
MFCD00013104
bis tetrabutylammonium dichromate, tbadc, tetrabutylammonium dichromate, bis tetrabutyl ammonium dichromate, acmc-1avk0, tetra-n-butylammonium dichromate, bis tetrabutylammonium ion chromate
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;tetrabutylazanium
Safety and Handling

Safety and Handling

GHS H Statement
H272-H350-H317
May intensify fire; oxidizer.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

SDS Documents