missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium chloride hydrate

Manufacturer:  Alfa Aesar™ 1934706

 View more versions of this product

Catalog No. AA1934706



Specifications

Specifications

Tetra-n-butylammonium chloride hydrate
73°C to 75°C
37451-68-6
MFCD00011635
FODWRUPJCKASBN-UHFFFAOYSA-M
tetrabutylazanium;chloride;hydrate
16212273
Crystalline
Hygroscopic
5g
C16H38ClNO
tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
CCCC[N+](CCCC)(CCCC)CCCC.O.[Cl-]
295.936
277.92 (Anhydrous)

Chemical Identifiers

37451-68-6
295.936
FODWRUPJCKASBN-UHFFFAOYSA-M
16212273
CCCC[N+](CCCC)(CCCC)CCCC.O.[Cl-]
C16H38ClNO
MFCD00011635
tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
tetrabutylazanium;chloride;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents