Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium borohydride, 98%

Chemical Identifiers

Chemical Name or Material Tetra-n-butylammonium borohydride
CAS 33725-74-5
Molecular Formula C16H40BN
Linear Formula [CH3(CH2)3]4NBH4
Formula Weight 257.31
MDL Number MFCD00012035
UN Number UN3131
Synonym Tetra-n-butylammonium tetrahydridoborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1749409

  SDS
alfa aesar™
A1749409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1749418

  SDS
alfa aesar™
A1749418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1749430

  SDS
alfa aesar™
A1749430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.31
Flash Point 140°C (284°F)
Health Hazard 1 Danger
Melting Point 125° to 131°C
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Quantity 10g
Molecular Formula C16H40BN
Synonym Tetra-n-butylammonium tetrahydridoborate
Assay 98%
CAS 33725-74-5
Linear Formula [CH3(CH2)3]4NBH4
MDL Number MFCD00012035
Chemical Name or Material Tetra-n-butylammonium borohydride
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
TSCA TSCA
UN Number UN3131
Health Hazard 2 GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.