missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-butylammonium acetate

Manufacturer:  Alfa Aesar™ 1927903

 View more versions of this product

Catalog No. AA1927903



Specifications

Specifications

Tetra-n-butylammonium acetate
White
1g
C18H39NO2
MFCD00043208
MCZDHTKJGDCTAE-UHFFFAOYSA-M
tetrabutylazanium;acetate
82707
Crystalline
Hygroscopic
114°C to 115°C
10534-59-5
[CH3(CH2)3]4N(O2CCH3)
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
CCCC[N+](CCCC)(CCCC)CCCC.CC(=O)[O-]
301.515
301.51

Chemical Identifiers

10534-59-5
301.515
MCZDHTKJGDCTAE-UHFFFAOYSA-M
82707
CCCC[N+](CCCC)(CCCC)CCCC.CC(=O)[O-]
C18H39NO2
MFCD00043208
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
tetrabutylazanium;acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents