Learn More
Tetra-n-butylammonium acetate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01927906
Description
Tetra-n-butylammonium acetate  is used as catalyst, curing agent, and initiator in the polymer industry. It is also used catalyst for the alkynylation of carbonyl compounds with trimethylsilylacetylenes. It is also used as PE/PP clarifying agent, epoxy polymerization initiator, surfactants, emulsifiers, and antistatic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-butylammonium acetate | |
114°C to 115°C | |
C18H39NO2 | |
MFCD00043208 | |
Hygroscopic | |
Soluble in acetonitrile (0.1g/mL). | |
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
301.52 | |
301.51 |
10534-59-5 | |
White | |
[CH3(CH2)3]4N(O2CCH3) | |
5 g | |
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
tetrabutylazanium;acetate | |
82707 | |
Crystalline |
Chemical Identifiers
10534-59-5 | |
301.52 | |
MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
82707 | |
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
C18H39NO2 | |
MFCD00043208 | |
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
tetrabutylazanium;acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 234-101-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only