missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Humboldt Test Tube Clamps

Easy to carry and pour

Manufacturer:  Humboldt HUMH8590

 View more versions of this product

Catalog No. 05-840



Description

Description

  • Stoddard-style clamps
  • Self-closing, rectangular jaw

Order Info

  • Jaw opening: up to 1 in. (25mm)
  • Length: 5 in. (12.7cm)

Specifications

Specifications

Test Tube Clamp
With finger grips
12.7 cm
Nickel Plated, Brass, Steel
5 in.
SDS
Documents
Product Certifications