Please login to your online account to display your discounted pricing

American Educational Products Test Tube Brush Bristles

Brush used to clean test tubes.

$12.25 - $18.00

Specifications

Color Natural
Product Type Test Tube Brush
For Use With (Application) For cleaning test tubes
View More Specs
Products
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity    

S30417D

 
american educational products
7-1027
Large 9.44 in. 24cm Dozen of 12 Each for $18.00

S30417F

 
american educational products
7-1025
Small 5.9 in. 15cm Dozen of 12 Each for $12.60

S30417E

 
american educational products
7-1026
Medium 7.87 in. 20cm Dozen of 12 Each for $12.25
Description & Specifications

Specifications

Color Natural
Product Type Test Tube Brush
For Use With (Application) For cleaning test tubes

  • Natural bristle
  • Available in small, medium and large size