missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TES, 0.2M buffer soln., pH 8.0

$88.36 - $138.47

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.229
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
IUPAC Name sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
SMILES C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61889AE View Documents Alfa Aesar™
J61889AE
100mL Each for $88.36
 
AAJ61889AK View Documents Alfa Aesar™
J61889AK
250mL Each for $138.47
 
Description

Description

Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

70331-82-7
251.229
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
C6H14NNaO6S
MFCD00065482
23702152
C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]

Specifications

70331-82-7
MFCD00065482
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
23702152
8.0
TES buffer soln.
C6H14NNaO6S
0.2M buffer soln.
C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]
251.229
Liquid
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA