missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TES, 0.2M buffer soln., pH 7.0

$84.96 - $133.08

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.229
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
IUPAC Name sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
SMILES C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63379AE View Documents Alfa Aesar™
J63379AE
100mL Each for $84.96
 
AAJ63379AK View Documents Alfa Aesar™
J63379AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

70331-82-7
251.229
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
C6H14NNaO6S
MFCD00065482
23702152
C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]

Specifications

70331-82-7
MFCD00065482
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
23702152
7.0
TES buffer soln.
C6H14NNaO6S
0.2M buffer soln.
C(CS(=O)(=O)[O-])NC(CO)(CO)CO.[Na+]
251.229
Liquid
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA