Please login to your online account to display your discounted pricing
Terumo™ Capiject™ Capillary Blood Collection Tubes
Single-piece, tapered design for easy collection and handling
$1,449.95 - $1,449.95
Specifications
Closure Included Yes
View More Specs
Products
Catalog Number Mfr. No. Applications Color Capacity (Metric) Price Quantity    
22-225-516
terumo™
TM
Used when a clotted sample is required; especially useful for blood banking or other laboratory procedures requiring serum Red 500μL Case of 1000 Each for $1,449.95
22-225-524
terumo™
TMG
Forms a barrier between serum and cells when the tube is centrifuged Cherry Red 500μL Case of 1000 Each for $1,449.95
22-225-540
terumo™
TMLH
Useful for many chemistry procedures that require heparinized whole blood or plasma Green 500μL Case of 1000 Each for $1,449.95
22-225-565
terumo™
TMPS
Optimal for procedures where glycolysis inhibitions are important Gray 250μL Case of 1000 Each for $1,449.95
22-245-856
terumo™
TMGA
Useful for preserving light-sensitive sample integrity Gold 500μL Case of 1000 Each for $1,449.95
Description & Specifications
Specifications
Closure Included Yes
  • Extra-wide, built-in collection lip for enhanced specimen flow
  • Color-coded fill lines and caps for volume accuracy and easy identification
  • Tube nests in ridged cap for convenient stand-alone placement
  • Lot number and expiration date printed on every tube
  • Silicone-coated anticoagulant tubes designed to reduce clotting
  • Olefin-based gel for clean separations
  • Certified for lead testing
  • Latex-free
Tube Functions:
  • Potassium EDTA: Provides a ratio of anti-coagulant to blood that is optimal for most procedures requiring whole blood or plasma
  • Lithium Heparin: Useful for many chemistry procedures that require heparinized whole blood or plasma
  • Sodium Fluoride/Potassium Oxalate: Optimal for procedures where glycolysis inhibitions are important
  • Gel Barrier: Forms a barrier between serum and cells when the tube is centrifuged
  • Amber-Colored Tube: Useful for preserving light-sensitive sample integrity
  • Plain Tube: Used when a clotted sample is required; especially useful for blood banking or other laboratory procedures requiring serum
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.