missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Nitrosobutane dimer, 98+%, Thermo Scientific Chemicals
$66.33 - $158.70
Chemical Identifiers
CAS | 6841-96-9 |
---|---|
Molecular Formula | C8H18N2O2 |
Molecular Weight (g/mol) | 174.244 |
MDL Number | MFCD00002065 |
InChI Key | PKKJRWITDTTZCL-UHFFFAOYSA-N |
Synonym | dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide |
PubChem CID | 44134826 |
IUPAC Name | 2-methyl-2-nitrosopropane |
SMILES | CC(C)(C)N=O.CC(C)(C)N=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL00462MD
|
Thermo Scientific Chemicals
L00462MD |
250 mg |
Each for $66.33
|
|
|||||
AAL0046203
|
Thermo Scientific Chemicals
L0046203 |
1 g |
Each for $158.70
|
|
|||||
Description
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.
Solubility
Soluble in 5% chloroform.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
6841-96-9 | |
174.244 | |
PKKJRWITDTTZCL-UHFFFAOYSA-N | |
44134826 | |
CC(C)(C)N=O.CC(C)(C)N=O |
C8H18N2O2 | |
MFCD00002065 | |
dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide | |
2-methyl-2-nitrosopropane |
Specifications
6841-96-9 | |
C8H18N2O2 | |
250 mg | |
dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide | |
PKKJRWITDTTZCL-UHFFFAOYSA-N | |
2-methyl-2-nitrosopropane | |
44134826 | |
≥98% |
88°C to 92°C (sublimation) | |
MFCD00002065 | |
Light sensitive | |
Soluble in 5% chloroform. | |
CC(C)(C)N=O.CC(C)(C)N=O | |
174.244 | |
174.24 | |
tert-Nitrosobutane dimer |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only