Please login to your online account to display your discounted pricing

tert-Butylurea 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC403700100

 View more versions of this product

Catalog No. AC403700100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 116.16
Quantity 10g
Color White
EINECS Number 214-257-3
TSCA TSCA
Melting Point 178° to 182°C
RTECS Number YS3680000
Chemical Name or Material tert-Butylurea
Name Note 98%
Molecular Formula C5H12N2O
Linear Formula (CH3)3CNHCONH2
MDL Number MFCD00025448
CAS 1118-12-3
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butylurea
Name Note 98%
CAS 1118-12-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H12N2O
Linear Formula (CH3)3CNHCONH2
Formula Weight 116.16
MDL Number MFCD00025448
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents