missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butylurea, 98%, ACROS Organics™
$570.38 - $570.38
Chemical Identifiers
CAS | 1118-12-3 |
---|---|
Molecular Formula | C5H12N2O |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00025448 |
InChI Key | JLEHSYHLHLHPAL-UHFFFAOYSA-N |
Synonym | 1,1-dimethylethyl urea, 1-tert-butylurea, n-tert-butylurea, t-butylurea, tert-butyl isourea, tert-butyl urea, tert-butyl-urea, urea, 1,1-dimethylethyl, urea, n-1,1-dimethylethyl, urea, tert-butyl |
PubChem CID | 14233 |
IUPAC Name | tert-butylurea |
SMILES | CC(C)(C)NC(=O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC403700250
|
Thermo Fisher Scientific
403700250 |
25g | Glass bottle |
Each for $570.38
|
|
Chemical Identifiers
1118-12-3 | |
116.16 | |
JLEHSYHLHLHPAL-UHFFFAOYSA-N | |
14233 | |
CC(C)(C)NC(=O)N |
C5H12N2O | |
MFCD00025448 | |
1,1-dimethylethyl urea, 1-tert-butylurea, n-tert-butylurea, t-butylurea, tert-butyl isourea, tert-butyl urea, tert-butyl-urea, urea, 1,1-dimethylethyl, urea, n-1,1-dimethylethyl, urea, tert-butyl | |
tert-butylurea |
Specifications
Authentic | |
White | |
25g | |
98% | |
(CH3)3CNHCONH2 | |
1,1-dimethylethyl urea, 1-tert-butylurea, n-tert-butylurea, t-butylurea, tert-butyl isourea, tert-butyl urea, tert-butyl-urea, urea, 1,1-dimethylethyl, urea, n-1,1-dimethylethyl, urea, tert-butyl | |
JLEHSYHLHLHPAL-UHFFFAOYSA-N | |
tert-butylurea | |
14233 | |
Crystals or Crystalline Powder | |
tert-Butylurea, 98% |
Glass bottle | |
178°C to 182°C | |
1118-12-3 | |
C5H12N2O | |
MFCD00025448 | |
Solubility in water: slightly soluble in water | |
CC(C)(C)NC(=O)N | |
116.16 | |
116.16 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 214-257-3
RTECSNumber : YS3680000
TSCA : TSCA