Please login to your online account to display your discounted pricing

tert-Butylsulfinamide, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426830010

 View more versions of this product

Catalog No. AC426830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystals
Assay ≥97.5%
Formula Weight 121.2
Quantity 1g
Color White to tan
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 146374-27-8
Molecular Formula C4H11NOS
Chemical Name or Material tert-Butylsulfinamide
Linear Formula (CH3)3CS(O)nH2
Synonym 2-Methyl-2-propanesulfinamide
Packaging Glass bottle

Specifications

Chemical Name or Material tert-Butylsulfinamide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H11NOS
Linear Formula (CH3)3CS(O)nH2
Formula Weight 121.2
Synonym 2-Methyl-2-propanesulfinamide