Learn More
tert-Butylmagnesium chloride, 1.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 331671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butylmagnesium chloride | |
1.7M solution in THF | |
C4H9ClMg | |
MFCD00000465 | |
0.9310g/mL | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
tert-butyl(chloro)magnesium | |
2724198 | |
19 to 21% (as BUMC) | |
0.931 | |
−17°C |
Brown | |
677-22-5,109-99-9 | |
(CH3)3CMgCl | |
04,63; 12,171 | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
CC(C)(C)[Mg]Cl | |
116.87 | |
116.87 | |
AcroSeal™ Glass bottle | |
100 mL | |
Liquid |
Chemical Identifiers
677-22-5 | |
116.87 | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
2724198 |
C4H9ClMg | |
MFCD00000465 | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
CC(C)(C)[Mg]Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
RUO – Research Use Only