Learn More
tert-Butylmagnesium chloride, 1.7M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 394308000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butylmagnesium chloride | |
0.8000g/mL | |
−40°C | |
C4H9ClMg | |
MFCD00000465 | |
0.8 | |
Solubility in water: reacts violently with water | |
CC(C)(C)[Mg]Cl | |
116.87 | |
116.87 |
677-22-5,60-29-7 | |
34.0°C | |
AcroSeal™ Glass Bottle | |
(CH3)3CMgCl | |
800 mL | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
magnesium;2-methylpropane;chloride | |
2724198 |
Chemical Identifiers
677-22-5 | |
116.87 | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
2724198 | |
CC(C)(C)[Mg]Cl |
C4H9ClMg | |
MFCD00000465 | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
magnesium;2-methylpropane;chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
In contact with water releases flammable gases which may ignite spontaneously.
Extremely flammable liquid and
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
EINECSNumber : 211-638-6
RUO – Research Use Only