Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyllithium, nominally 1.5M in n-pentane, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material tert-Butyllithium
CAS 594-19-4
Linear Formula (CH3)3CLi
Formula Weight 64.06
MDL Number MFCD00008795
UN Number UN3394
Beilstein 3587204
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41251AD

 
alfa aesar™
41251AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 64.06
Chemical Name or Material tert-Butyllithium
Concentration Nominally 1.5M in n-pentane
TSCA TSCA
Physical Form Liquid
Linear Formula (CH3)3CLi
MDL Number MFCD00008795
CAS 594-19-4
Quantity 50mL
UN Number UN3394
Beilstein 3587204
Density 0.652g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H260-H252-H304-H314-H336
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Cau
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless to pale yellow
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H260-H252-H304-H314-H336
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Cau

  • Packaged under Argon