missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylhydroquinone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 150825000

 View more versions of this product

Catalog No. AC150825000



Description

Description

Specifications

Specifications

tert-Butylhydroquinone
Authentic
295.0°C
125.0°C to 130.0°C
97%
96% min. (GC)
(CH3)3CC6H3(OH)2
06, IV, 6013
Solubility in water: 1g/100mL (25 c). Other solubilities: soluble in acetone (190/100g at 25 c),,soluble in ethanol (120/100g at 25 c),,soluble in ethyl acetate (100g/100g at 25 c),,soluble in dmso,diethyl ether;,insoluble in benzene,toluene
CC(C)(C)C1=C(C=CC(=C1)O)O
166.22
CHEBI:78886
Crystalline Powder
171°C
Plastic bottle
Black-Tan to White
500g
1948-33-0
C10H14O2
MFCD00002344
2-t-butylhydroquinone, 2-tert-butyl-1,4-benzenediol, 2-tert-butylhydroquinone, mono-tert-butylhydroquinone, mtbhq, sustane, t-butyl hydroquinone, t-butylhydroquinone, tbhq, tert-butylhydroquinone
BGNXCDMCOKJUMV-UHFFFAOYSA-N
2-tert-butylbenzene-1,4-diol
16043
166.22
97%

Chemical Identifiers

1948-33-0
166.22
BGNXCDMCOKJUMV-UHFFFAOYSA-N
16043
2-tert-butylbenzene-1,4-diol
C10H14O2
MFCD00002344
2-t-butylhydroquinone, 2-tert-butyl-1,4-benzenediol, 2-tert-butylhydroquinone, mono-tert-butylhydroquinone, mtbhq, sustane, t-butyl hydroquinone, t-butylhydroquinone, tbhq, tert-butylhydroquinone
CHEBI:78886
CC(C)(C)C1=C(C=CC(=C1)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-752-2

RTECSNumber : MX4375000

TSCA : TSCA

SDS Documents