missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylhydrazine hydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 7400-27-3
Molecular Formula C4H13ClN2
Molecular Weight (g/mol) 124.61
MDL Number MFCD00012947
InChI Key DDPWVABNMBRBFI-UHFFFAOYSA-N
Synonym 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, hydrazine, 1,1-dimethylethyl-, monohydrochloride, t-bunhnh2.hcl, t-butylhydrazine hydrochloride, t-butylhydrazine.hcl, tert-butylhydrazine hydrochloride, tert-butylhydrazine monohydrochloride, tert-butylhydrazinium chloride
PubChem CID 81889
IUPAC Name tert-butylhydrazine;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)NN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC187081000
SDS View Documents Promotion Details
Thermo Fisher Scientific
187081000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7400-27-3
124.61
DDPWVABNMBRBFI-UHFFFAOYSA-N
81889
[H+].[Cl-].CC(C)(C)NN
C4H13ClN2
MFCD00012947
1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, hydrazine, 1,1-dimethylethyl-, monohydrochloride, t-bunhnh2.hcl, t-butylhydrazine hydrochloride, t-butylhydrazine.hcl, tert-butylhydrazine hydrochloride, tert-butylhydrazine monohydrochloride, tert-butylhydrazinium chloride
tert-butylhydrazine;hydrochloride

Specifications

7400-27-3
190°C to 194°C
C4H13ClN2
(CH3)3CNHNH2·HCl
Glass bottle
12,87
1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, hydrazine, 1,1-dimethylethyl-, monohydrochloride, t-bunhnh2.hcl, t-butylhydrazine hydrochloride, t-butylhydrazine.hcl, tert-butylhydrazine hydrochloride, tert-butylhydrazine monohydrochloride, tert-butylhydrazinium chloride
[H+].[Cl-].CC(C)(C)NN
124.61
124.61
98%
White
98%
Authentic
MFCD00012947
04, III, 1734
100g
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine;hydrochloride
81889
Crystals or Crystalline Powder
tert-Butylhydrazine hydrochloride, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-001-6

RTECSNumber : MV2841000

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only