missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyldimethylsilyl Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T152525G

 View more versions of this product

Catalog No. T152525G



Specifications

Specifications

tert-Butyldimethylsilyl Trifluoromethanesulfonate
25g
C7H15F3O3SSi
2924
WLLIXJBWWFGEHT-UHFFFAOYSA-N
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
123564
Clear Liquid at 20°C
Yellow
69739-34-0
MFCD00000405
tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
264.334
264.33
≥98.0% (T)

Chemical Identifiers

69739-34-0
264.334
WLLIXJBWWFGEHT-UHFFFAOYSA-N
123564
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
C7H15F3O3SSi
MFCD00000405
tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents