Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyldimethylchlorosilane, 50% w/w in toluene

Chemical Identifiers

Chemical Name or Material tert-Butyldimethylchlorosilane
CAS 18162-48-6
Molecular Formula C6H15ClSi
Linear Formula (CH3)3CSi(CH3)2Cl
Formula Weight 150.73
MDL Number MFCD00000501
UN Number UN2924
Synonym tert-Butylchlorodimethylsilane; tert-Butyldimethylsilyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB212860B

 
alfa aesar™
B212860B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2128618

 
alfa aesar™
B2128618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2128630

 
alfa aesar™
B2128630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.73
TSCA TSCA
Flash Point 7°C (45°F)
Health Hazard 2 GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Chemical Name or Material tert-Butyldimethylchlorosilane
Odor Characteristic
Molecular Formula C6H15ClSi
Linear Formula (CH3)3CSi(CH3)2Cl
MDL Number MFCD00000501
CAS 18162-48-6
UN Number UN2924
Synonym tert-Butylchlorodimethylsilane; tert-Butyldimethylsilyl chloride
Quantity 1000g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Concentration 50% w/w in toluene
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.