Learn More
tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 183931000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butylchlorodimethylsilane | |
85.0°C to 91.0°C | |
0.8700g/mL | |
22°C | |
97.5% min. (GC) | |
C6H15ClSi | |
MFCD00000501 | |
2925 | |
0.87 | |
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
tert-butyl-chloro-dimethylsilane | |
28928 | |
150.73 | |
Crystals |
18162-48-6 | |
Colorless to White | |
125.0°C | |
Authentic | |
Glass bottle | |
(CH3)3CSi(CH3)2Cl | |
100 g | |
04,176; 05,74; 06,78; 08,59; 09,77; 10,62; 13,87; 17,5 | |
15, 1567 | |
Solubility in water: reacts. Other solubilities: reacts with alcohols, organic acids and amines, soluble in ethers and hydrocarbons | |
CC(C)(C)[Si](C)(C)Cl | |
150.73 | |
CHEBI:85071 | |
98% |
Chemical Identifiers
18162-48-6 | |
150.73 | |
BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
28928 | |
tert-butyl-chloro-dimethylsilane |
C6H15ClSi | |
MFCD00000501 | |
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
CHEBI:85071 | |
CC(C)(C)[Si](C)(C)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha
GHS Signal Word: Danger
EINECSNumber : 242-042-4
RUO – Research Use Only