Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butylbenzene, 99%

Chemical Identifiers

Chemical Name or Material tert-Butylbenzene
CAS 98-06-6
Molecular Formula C10H14
Formula Weight 134.22
MDL Number MFCD00008816
UN Number UN2709
Beilstein 1421537
Merck Index 14,1551
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA12378AK

  SDS
alfa aesar™
A12378AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.22
Chemical Name or Material tert-Butylbenzene
TSCA TSCA
Assay 99%
Molecular Formula C10H14
MDL Number MFCD00008816
CAS 98-06-6
UN Number UN2709
Beilstein 1421537
Merck Index 14,1551
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 168° to 169°C
Melting Point −58°C
Flash Point 34°C (93°F)
Odor Characteristic
Quantity 250mL
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
RTECS Number CY9120000

Applications
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

Solubility
Insoluble in water(0.03g/100 mL at 20°C) .

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.