missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylamine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107820050

 View more versions of this product

Catalog No. AC107820050


 

Specifications Description & Specifications

Specifications

tert-Butylamine
Authentic
≥98.5%
Glass Bottle
1.3770 to 1.3790 (20°C, 589nm)
73.13
01,84; 02,43; 10,62; 11,138
Clear
75-64-9
Liquid
99%
C4H11N
(CH3)3CNH2
MFCD00008050
1% max.
5mL

Chemical Identifiers

99%
(CH3)3CNH2
MFCD00008050
≥98.5%
C4H11N
73.13
tert-Butylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger