Please login to your online account to display your discounted pricing

tert-Butyl trimethylacetate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material tert-Butyl trimethylacetate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H18O2
Linear Formula (CH3)3CCO2C(CH3)3
Formula Weight 158.24
MDL Number MFCD01861974
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387620010

  SDS
thermo fisher scientific
AC387620010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid, Darkening During Storage
Formula Weight 158.24
Color Clear colorless
Quantity 1g
Density 0.8130g/mL
Chemical Name or Material tert-Butyl trimethylacetate
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CH3)3CCO2C(CH3)3
Packaging Glass bottle
MDL Number MFCD01861974
CAS 16474-43-4
Refractive Index 1.3910 to 1.3930 (20°C, 589nm)
Molecular Formula C9H18O2