Please login to your online account to display your discounted pricing

tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  AW01051DA

 View more versions of this product

Catalog No. AW01051DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.25
Quantity 1g
CAS 57260-71-6
Assay Percent Range 97%
Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Molecular Formula C9H18N2O2
Synonym N-BOC-Piperazine

Specifications

Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Assay Percent Range 97%
Molecular Formula C9H18N2O2
Formula Weight 186.25
Synonym N-BOC-Piperazine