Please login to your online account to display your discounted pricing

tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Assay Percent Range 97%
Molecular Formula C9H18N2O2
Formula Weight 186.25
Synonym N-BOC-Piperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AW01051DA

 
fisher scientific company
AW01051DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AW01051EA

 
fisher scientific company
AW01051EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.25
Quantity 1g
Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Assay Percent Range 97%
Molecular Formula C9H18N2O2
CAS 57260-71-6
Synonym N-BOC-Piperazine