Please login to your online account to display your discounted pricing

tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  AW01051EA

 View more versions of this product

Catalog No. AW01051EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.25
Quantity 10g
CAS 57260-71-6
Synonym N-BOC-Piperazine
Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Assay Percent Range 97%
Molecular Formula C9H18N2O2

Specifications

Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Assay Percent Range 97%
Molecular Formula C9H18N2O2
Formula Weight 186.25
Synonym N-BOC-Piperazine