missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 386570010

 View more versions of this product

Catalog No. AC386570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

tert-Butyl (S)-(-)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
1.0600g/mL
108°C
94% min. (GC)
C11H19NO4
MFCD00209557
1.06
-85° to -95° (20°C, 589nm) (c=1, CHCl3)
PNJXYVJNOCLJLJ-MRVPVSSYSA-N
tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
179824
95%
102308-32-7
83°C to 88°C (1.0mmHg)
Authentic
Glass bottle
1.4450 to 1.4490 (20°C, 589nm)
1g
15,4404
s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
229.28
229.28

Chemical Identifiers

102308-32-7
229.28
PNJXYVJNOCLJLJ-MRVPVSSYSA-N
179824
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
C11H19NO4
MFCD00209557
s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only